Learn More
Cayman Chemical Alamethicin, Purity: ≥98% (mixture of isoforms), CAS: 27061-78-5, 10 mg
SDP
Catalog No. 502951518
Encompass_Preferred
A membrane active peptide antibiotic of the peptaibol class that strongly induces helical peptide structures and forms voltage-gated ion channels in the lipid bilayers of cell membranes; often used to study ion channel assembly, voltage gating, and peptide-membrane interactions as well as to induce defense responses in plants
Catalog No. 50-295-1518 Supplier Cayman Chemical Supplier No. 1142510 MG
A membrane active peptide antibiotic of the peptaibol class that strongly induces helical peptide structures and forms voltage-gated ion channels in the lipid bilayers of cell membranes; often used to study ion channel assembly, voltage gating, and peptide-membrane interactions as well as to induce defense responses in plants