Learn More
Enzo Life Sciences Alamethicin (25mg). CAS: 27061-78-5
SDP
Catalog No. 502010226
Encompass_Preferred
Mixture of alamethicin homologs. Membrane permeabilizing antibiotic. Applications include increasing [32P] incorporation into phosphatidylinositol 4-phosphate and permeabilization of sarcoplasmic reticulum vesicles. Causes fusion of lipid vesicles. Alternative name: U-22324. Purity: ≥98% (TLC). Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: +4°C.
Catalog No. 50-201-0226 Supplier Enzo Life Sciences Supplier No. BMLA1500025
Mixture of alamethicin homologs. Membrane permeabilizing antibiotic. Applications include increasing [32P] incorporation into phosphatidylinositol 4-phosphate and permeabilization of sarcoplasmic reticulum vesicles. Causes fusion of lipid vesicles. Alternative name: U-22324. Purity: ≥98% (TLC). Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: +4°C.