Learn More
Medchemexpress LLC Alamethicin | 27061-78-5 | 10 MG
SDP
Catalog No. 5000420186
Encompass_Preferred
Alamethicin is a linear 20-amino acid antibiotic that induces voltage-gated conductance in model and cell membranes. It exhibits antimicrobial activity against Gram-positive bacteria, but not Gram-negative bacteria. Alamethicin can form an amphipathic α-helical structure in biological membranes.
  • Induces voltage-gated conductance in model and cell membranes.
  • Exhibits antimicrobial activity against gram-positive bacteria.
  • Forms an amphipathic α-helical structure in biological membranes.
  • Possesses an unusual primary sequence with non-standard amino acids and modifications.
  • Forms barrel-stave pores in model lipid multilayers.
Catalog No. 50-004-20186 Supplier Medchemexpress LLC Supplier No. HYN670810MG
Alamethicin is a linear 20-amino acid antibiotic that induces voltage-gated conductance in model and cell membranes. It exhibits antimicrobial activity against Gram-positive bacteria, but not Gram-negative bacteria. Alamethicin can form an amphipathic α-helical structure in biological membranes.
  • Induces voltage-gated conductance in model and cell membranes.
  • Exhibits antimicrobial activity against gram-positive bacteria.
  • Forms an amphipathic α-helical structure in biological membranes.
  • Possesses an unusual primary sequence with non-standard amino acids and modifications.
  • Forms barrel-stave pores in model lipid multilayers.