Learn More
Medchemexpress LLC Alamethicin | 27061-78-5 | 5 MG
SDP
Catalog No. 5000420183
Encompass_Preferred
Alamethicin is a linear 20-amino acid antibiotic that induces voltage-gated conductance in model and cell membranes. It exhibits antimicrobial activity against Gram-positive bacteria and forms amphipathic α-helical structures in biological membranes. Its unusual primary sequence includes non-standard amino acids and C- and N-terminal modifications.
  • Induces voltage-gated conductance in membranes
  • Exhibits antimicrobial activity against gram-positive bacteria
  • Forms amphipathic α-helical structures in biological membranes
  • Molecular weight of 1964.34
  • Appears as a white to yellow solid
  • Forms barrel-stave pores in model lipid multilayers composed of phosphatidylcholine
Catalog No. 50-004-20183 Supplier Medchemexpress LLC Supplier No. HYN67085MG
Alamethicin is a linear 20-amino acid antibiotic that induces voltage-gated conductance in model and cell membranes. It exhibits antimicrobial activity against Gram-positive bacteria and forms amphipathic α-helical structures in biological membranes. Its unusual primary sequence includes non-standard amino acids and C- and N-terminal modifications.
  • Induces voltage-gated conductance in membranes
  • Exhibits antimicrobial activity against gram-positive bacteria
  • Forms amphipathic α-helical structures in biological membranes
  • Molecular weight of 1964.34
  • Appears as a white to yellow solid
  • Forms barrel-stave pores in model lipid multilayers composed of phosphatidylcholine