Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alamethicin, ≥50%, MP Biomedicals™

Catalog No. ICN15900905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15900905 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15900905 Supplier MP Biomedicals, Inc Supplier No. 0215900905
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59588-86-2
Molecular Formula C92H150N22O25
Molecular Weight (g/mol) 1964.341
InChI Key LGHSQOCGTJHDIL-SLKIUSOBSA-N
Synonym U-22324 Antibiotic
PubChem CID 53229968
SMILES CC(C)CC(C(=O)NC(C)(C)C(=O)N1CCCC1C(=O)NC(C(C)C)C(=O)NC(C)(C)C(=O)NC(C)(C)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CC2=CC=CC=C2)CO)NC(=O)CNC(=O)C(C)(C)NC(=O)C(C(C)C)NC(=O)C(C)(C)NC(=O)C(CCC(=O)N)NC(=O)C(C)NC(=O)C(C)(C)NC(=O)C(C)NC(=O)C(C)(C)NC(=O)C3CCCN3C

Specifications

Type Alamethicin
Physical Form Powder
Color Tan
Quantity 5 mg
Assay Percent Range ≥50%
Formula Weight 1964.35
Chemical Name or Material Alamethicin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.