Learn More
Enzo Life Sciences Alamethicin (5 mg) CAS: 27061-78-5
SDP
Catalog No. NC1229980
Encompass_Preferred
Antibiotic that increases membrane permeability. Mixture of alamethicin homologs. Membrane permeabilizing antibiotic. Applications include increasing [32P] incorporation into phosphatidylinositol 4-phosphate and permeabilization of sarcoplasmic reticulum vesicles. Causes fusion of lipid vesicles. Alternative Name: U-22324. Formula: C92H150N22O25. MW: 1964.3. CAS: 27061-78-5. Purity: ≥98% (TLC). Sequence Ac-2-MeAla-L-Pro-2-MeAla-L-Ala-2-MeAla-L-Ala-L-Gln-2-MeAla-L-Val-2-MeAla-Gly-L-Leu-2-MeAla-L-Pro-L-Val-2-MeAla-2-MeAla-L-Glu-L-Gln-L-phenylalaninol. Appearance: White to off-white solid. Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: +4°C. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at 0-4°C. Stock solutions are stable for up to 3 months at -20°C.
Catalog No. NC1229980 Supplier Enzo Life Sciences Supplier No. BMLA1500005
Antibiotic that increases membrane permeability. Mixture of alamethicin homologs. Membrane permeabilizing antibiotic. Applications include increasing [32P] incorporation into phosphatidylinositol 4-phosphate and permeabilization of sarcoplasmic reticulum vesicles. Causes fusion of lipid vesicles. Alternative Name: U-22324. Formula: C92H150N22O25. MW: 1964.3. CAS: 27061-78-5. Purity: ≥98% (TLC). Sequence Ac-2-MeAla-L-Pro-2-MeAla-L-Ala-2-MeAla-L-Ala-L-Gln-2-MeAla-L-Val-2-MeAla-Gly-L-Leu-2-MeAla-L-Pro-L-Val-2-MeAla-2-MeAla-L-Glu-L-Gln-L-phenylalaninol. Appearance: White to off-white solid. Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: +4°C. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at 0-4°C. Stock solutions are stable for up to 3 months at -20°C.