Learn More
Cayman Chemical AlamethIcIn F50 10mg
SDP
Catalog No. 502993957
Encompass_Preferred
A neutral, linear peptaibol isolated from Trichoderma sp.; induces formation of voltage-dependent ion channels in lipid membranes; acts as a lytic (IC50 = 54 and 80 μM for rat mast cells and mouse spleen lymphocytes); exhibits antibacterial activity against mollicutes (MICs = 1.56-12.5 μM)
Catalog No. 50-299-3957 Supplier Cayman Chemical Supplier No. 2314110 MG
A neutral, linear peptaibol isolated from Trichoderma sp.; induces formation of voltage-dependent ion channels in lipid membranes; acts as a lytic (IC50 = 54 and 80 μM for rat mast cells and mouse spleen lymphocytes); exhibits antibacterial activity against mollicutes (MICs = 1.56-12.5 μM)