Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Alane N,N-Dimethylethylamine Complex Solution

Catalog No. 400386100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-038-6100ML 100 mL
40-038-650ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 40-038-6100ML Supplier Sigma Aldrich Supplier No. 400386100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of aluminum nanoparticlesFluorination reactionsAluminum metal-organic chemical vapor deposition for the growth of aluminum filmsLigand-exchange reactionsHydroalumination of nitriles and isonitriles

Specifications

CAS 124330-23-0
Density 0.837 g/mL at 25°C
Molecular Formula C4H14AlN
Linear Formula C2H5N(CH3)2 -+ AlH3
MDL Number MFCD00191709
Quantity 100 mL
Synonym (Ethyldimethylamine)trihydroaluminum; Dimethylethylamine alane
Molecular Weight (g/mol) 103.14 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.