Learn More
Medchemexpress LLC β-Alanine-13C3,15N | 285978-07-6 | 99.99% | 93.06 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0230S1MG
Encompass_Preferred
β-Alanine-13C3,15N is the 13C- and 15N-labeled β-Alanine. β-Alanine is a non-essential amino acid that is shown to be metabolized into carnosine, which functions as an intracellular buffer. This product is for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-31050