Learn More
Medchemexpress LLC Alanine-2,3,3,3-d4 | 53795-92-9 | MFCD00084108 | 99.8% | 93.12 | C3H3D4NO2 | 100 MG
SDP
Catalog No. 5000281284
Encompass_Preferred
Alanine-2,3,3,3-d4 is a deuterium-labeled alanine used as a stable-isotope tracer and internal standard for research and analytical workflows. It supports accurate quantitation and method development in biochemical and pharmacokinetic studies, dissolves readily in water, and is supplied with recommended storage conditions for long-term stability.
  • Deuterium-labeled alanine for tracer and internal standard use.
  • High purity (≈99.8%) suitable for quantitative analysis.
  • Soluble in water (≥ 50 mg/mL) for straightforward sample preparation.
  • Stable as a powder under frozen storage for long-term use.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
Catalog No. 50-002-81284 Supplier Medchemexpress LLC Supplier No. HYW244382100MG
Alanine-2,3,3,3-d4 is a deuterium-labeled alanine used as a stable-isotope tracer and internal standard for research and analytical workflows. It supports accurate quantitation and method development in biochemical and pharmacokinetic studies, dissolves readily in water, and is supplied with recommended storage conditions for long-term stability.
  • Deuterium-labeled alanine for tracer and internal standard use.
  • High purity (≈99.8%) suitable for quantitative analysis.
  • Soluble in water (≥ 50 mg/mL) for straightforward sample preparation.
  • Stable as a powder under frozen storage for long-term use.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.