Learn More
Medchemexpress LLC Alanine-2,3,3,3-d4 | 53795-92-9 | MFCD00084108 | 99.8% | 93.12 | C3H3D4NO2 | 25 MG
SDP
Catalog No. 5000280849
Encompass_Preferred
Alanine-2,3,3,3-d4 is a deuterium-labeled stable isotope of alanine intended for research use. It is suitable for use as an internal standard in analytical workflows, and for metabolic tracing and isotope labeling studies. The material is supplied with supporting documentation including a datasheet, certificate of analysis, and safety data sheet, and is not intended for human use.
  • Deuterium-labeled alanine for isotope tracing and analytical standards.
  • High isotopic and chemical purity (99.8%).
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR.
  • Available in small measured quantities for research workflows.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-80849 Supplier Medchemexpress LLC Supplier No. HYW24438225MG
Alanine-2,3,3,3-d4 is a deuterium-labeled stable isotope of alanine intended for research use. It is suitable for use as an internal standard in analytical workflows, and for metabolic tracing and isotope labeling studies. The material is supplied with supporting documentation including a datasheet, certificate of analysis, and safety data sheet, and is not intended for human use.
  • Deuterium-labeled alanine for isotope tracing and analytical standards.
  • High isotopic and chemical purity (99.8%).
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR.
  • Available in small measured quantities for research workflows.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.