Learn More
Medchemexpress LLC Alanine-2,3,3,3-d4 | 53795-92-9 | MFCD00084108 | 99.8% | 93.12 | C3H3D4NO2 | 5 MG
SDP
Catalog No. 5000280695
Encompass_Preferred
Alanine-2,3,3,3-d4 is the deuterium-labeled analogue of alanine supplied for research use as a stable isotope tracer or internal standard. It is intended for quantitative analysis by NMR, GC-MS, and LC-MS. Chemical properties include purity 99.81%, formula C3H3D4NO2, molecular weight 93.12, and CAS number 53795-92-9.
  • Deuterium-labeled analogue for isotope tracing.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • High purity for accurate quantitation.
  • Available in small mg-scale quantities for laboratory research.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-80695 Supplier Medchemexpress LLC Supplier No. HYW2443825MG
Alanine-2,3,3,3-d4 is the deuterium-labeled analogue of alanine supplied for research use as a stable isotope tracer or internal standard. It is intended for quantitative analysis by NMR, GC-MS, and LC-MS. Chemical properties include purity 99.81%, formula C3H3D4NO2, molecular weight 93.12, and CAS number 53795-92-9.
  • Deuterium-labeled analogue for isotope tracing.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • High purity for accurate quantitation.
  • Available in small mg-scale quantities for laboratory research.
  • For research use only; not for human or veterinary use.