Learn More
Medchemexpress LLC Alanine-2,3,3,3-d4 | 53795-92-9 | MFCD00084108 | 99.8% | 93.12 g/mol | C3H3D4NO2 | 10 MG
SDP
Catalog No. 5000282574
Encompass_Preferred
Alanine-2,3,3,3-d4 is a deuterium-labeled form of alanine used as a stable-isotope tracer and internal standard for quantitative analysis. It is intended for analytical workflows in which accurate quantitation and tracing are required, and is supplied in milligram pack sizes suitable for NMR, GC-MS, and LC-MS applications.
  • Deuterium-labeled for isotope tracing and internal standardization.
  • High purity (99.8%) for accurate quantitation.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in multiple small milligram pack sizes for analytical use.
  • Molecular weight 93.12 g/mol and CAS 53795-92-9 for unambiguous identification.
Catalog No. 50-002-82574 Supplier Medchemexpress LLC Supplier No. HYW24438210MG
Alanine-2,3,3,3-d4 is a deuterium-labeled form of alanine used as a stable-isotope tracer and internal standard for quantitative analysis. It is intended for analytical workflows in which accurate quantitation and tracing are required, and is supplied in milligram pack sizes suitable for NMR, GC-MS, and LC-MS applications.
  • Deuterium-labeled for isotope tracing and internal standardization.
  • High purity (99.8%) for accurate quantitation.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in multiple small milligram pack sizes for analytical use.
  • Molecular weight 93.12 g/mol and CAS 53795-92-9 for unambiguous identification.