Learn More
Medchemexpress LLC Alanine-2,3,3,3-d4 | 53795-92-9 | MFCD00084108 | 99.8% | 93.12 g/mol | C3H3D4NO2 | 50 MG
SDP
Catalog No. 5000281287
Encompass_Preferred
Alanine-2,3,3,3-d4 is a deuterium-labeled alanine used as a stable isotope standard for analytical and biochemical applications. Supplied as a powder in multiple package sizes, it is suitable for mass spectrometry, nuclear magnetic resonance (NMR), isotope-dilution quantitation, and metabolic tracing. High isotopic and chemical purity, together with available documentation, supports reproducible analytical workflows.
  • High isotopic purity (deuterium-labeled) for accurate internal standard use.
  • High chemical purity for reliable analytical results.
  • Available in small, research-scale quantities to minimize sample waste.
  • Supplied with datasheet, certificate of analysis, and safety data sheet for quality and safety documentation.
  • Stable under recommended storage conditions to preserve compound integrity.
Catalog No. 50-002-81287 Supplier Medchemexpress LLC Supplier No. HYW24438250MG
Alanine-2,3,3,3-d4 is a deuterium-labeled alanine used as a stable isotope standard for analytical and biochemical applications. Supplied as a powder in multiple package sizes, it is suitable for mass spectrometry, nuclear magnetic resonance (NMR), isotope-dilution quantitation, and metabolic tracing. High isotopic and chemical purity, together with available documentation, supports reproducible analytical workflows.
  • High isotopic purity (deuterium-labeled) for accurate internal standard use.
  • High chemical purity for reliable analytical results.
  • Available in small, research-scale quantities to minimize sample waste.
  • Supplied with datasheet, certificate of analysis, and safety data sheet for quality and safety documentation.
  • Stable under recommended storage conditions to preserve compound integrity.