Learn More
Medchemexpress LLC HY-N0230 500mg Medchemexpress, β-Alanine CAS:107-95-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0230500MG
Encompass_Preferred
Medchemexpress, HY-N0230 500mg β-Alanine CAS:107-95-9 β-Alanine is a non-essential amino acid that is shown to be metabolized into carnosine, which functions as an intracellular buffer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2779