Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Alanine, 99+%, Thermo Scientific™

Catalog No. AC219590010
Change view
Click to view available options
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC219590010 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC219590010 Supplier Thermo Scientific Chemicals Supplier No. 219590010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 107-95-9
Ammonium 200ppm max.
CAS Min % 99.0
CAS Max % 100.0
Chlorine (Cl) 400ppm max.
Foreign Amino Acids (TLC) Passes Test (not detectable on 10μgspotting)
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 99+%
Molecular Formula C3H7NO2
Linear Formula H2NCH2CH2CO2H
Infrared Spectrum Authentic
MDL Number MFCD00008200
Iron (Fe) 30ppm max.
Beilstein 04, 401
Fieser 01,16; 05,6; 06,10; 17,3
Loss on Drying 0.2% max. (105°C, 3 hrs)
Merck Index 15, 198
Synonym beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
InChI Key UCMIRNVEIXFBKS-UHFFFAOYSA-N
SMILES NCCC(O)=O
IUPAC Name 3-aminopropanoic acid
Molecular Weight (g/mol) 89.09
PubChem CID 239
ChEBI CHEBI:16958
Sulfate 480ppm max.
Formula Weight 89.09
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Percent Purity 99.0 to 101.0% (on dry substance)
Sulfated Ash 0.2% max.
Quantity 1kg
Chemical Name or Material β-Alanine, 99+%
Show More Show Less
EINECSNumber 203-536-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.