Learn More
Medchemexpress LLC Alanine dehydrogenase, expressed in E. coli | 9029-06-5 | ≥15 U/mg solid | 10 MG
SDP
Catalog No. 5000398634
Encompass_Preferred
Alanine dehydrogenase, expressed in E. coli, is a microbial enzyme that catalyzes a reversible conversion of L-alanine to pyruvate. It has an EC Number of 1.4.1.1 and an enzyme activity of ≥15 U/mg solid. One unit corresponds to the amount of enzyme which converts 1 μmol L-alanine per minute at pH 10.0 and 30°C. This product appears as a solid, white to off-white in color and is for research use only.
  • Catalyzes reversible conversion of L-alanine to pyruvate
  • EC Number: 1.4.1.1
  • Enzyme activity: ≥15 U/mg solid
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-003-98634 Supplier Medchemexpress LLC Supplier No. HYP285410MG
Alanine dehydrogenase, expressed in E. coli, is a microbial enzyme that catalyzes a reversible conversion of L-alanine to pyruvate. It has an EC Number of 1.4.1.1 and an enzyme activity of ≥15 U/mg solid. One unit corresponds to the amount of enzyme which converts 1 μmol L-alanine per minute at pH 10.0 and 30°C. This product appears as a solid, white to off-white in color and is for research use only.
  • Catalyzes reversible conversion of L-alanine to pyruvate
  • EC Number: 1.4.1.1
  • Enzyme activity: ≥15 U/mg solid
  • White to off-white solid appearance
  • For research use only