Learn More
Apexbio Technology LLC β-Alanine methyl ester hydrochloride 3196-73-4 1g
SDP
Catalog No. 5000094638
Encompass_Preferred
beta-Alanine methyl ester hydrochloride is a synthetic intermediate used in the preparation of bipyridine-based ligands for metal ion chelation It is designed to facilitate the formation of coordination complexes thereby enabling studies on metal ion-biological interactions and enzyme catalytic mechanisms Based on these properties beta-Alanine methyl ester hydrochloride holds research potential in biological labeling methodologies drug delivery systems and the investigation of metal ion mechanisms and biocompatibility within biomedical research fields
Catalog No. 50-000-94638 Supplier Apexbio Technology LLC Supplier No. C71111000
beta-Alanine methyl ester hydrochloride is a synthetic intermediate used in the preparation of bipyridine-based ligands for metal ion chelation It is designed to facilitate the formation of coordination complexes thereby enabling studies on metal ion-biological interactions and enzyme catalytic mechanisms Based on these properties beta-Alanine methyl ester hydrochloride holds research potential in biological labeling methodologies drug delivery systems and the investigation of metal ion mechanisms and biocompatibility within biomedical research fields