Learn More
Medchemexpress LLC β-alanine, N-[4-[[7-[(aminoiminomethyl)amino]-1-oxoheptyl]amino]-3-hydroxy-1-oxobutyl]- | 140686-92-6 | ≥98.0% | 359.42 g/mol | C15H29N5O | 100 MG
SDP
Catalog No. 5000211361
Encompass_Preferred
N-563 is a deoxyspergualin-mimetic research compound reported to stimulate immune responses and enhance resistance to Candida albicans in mice. It is provided as a solid research reagent (CAS 140686-92-6) with a reported molecular weight of 359.42 g/mol and typical purity ≥98%.
  • Deoxyspergualin mimetic that stimulates immune responses.
  • Enhances resistance to Candida albicans in murine models.
  • Suitable for in vitro and in vivo immunology studies.
  • Reported high purity for reliable experimental results.
  • Provided as a solid, typically soluble in DMSO for assay preparation.
Catalog No. 50-002-11361 Supplier Medchemexpress LLC Supplier No. HY100751100MG
N-563 is a deoxyspergualin-mimetic research compound reported to stimulate immune responses and enhance resistance to Candida albicans in mice. It is provided as a solid research reagent (CAS 140686-92-6) with a reported molecular weight of 359.42 g/mol and typical purity ≥98%.
  • Deoxyspergualin mimetic that stimulates immune responses.
  • Enhances resistance to Candida albicans in murine models.
  • Suitable for in vitro and in vivo immunology studies.
  • Reported high purity for reliable experimental results.
  • Provided as a solid, typically soluble in DMSO for assay preparation.