Learn More
Medchemexpress LLC Alanine, n-[[5-chloro-2-(3-cyanophenyl)methoxy]-4-(2-methyl-biphenyl-3-yl)methoxy]... | 2377916-66-8 | 98.3% | C33H31ClN2O4 | 10MG
SDP
Catalog No. 5000261267
Encompass_Preferred
PD-1/PD-L1-IN-NP19 is a small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction, reported to block the interaction with an IC50 of 12.5 nM. Supplied as a purified solid for research use, it is intended for in vitro and preclinical studies where modulation of the immune microenvironment is required.
Catalog No. 50-002-61267 Supplier Medchemexpress LLC Supplier No. HY13134710MG
PD-1/PD-L1-IN-NP19 is a small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction, reported to block the interaction with an IC50 of 12.5 nM. Supplied as a purified solid for research use, it is intended for in vitro and preclinical studies where modulation of the immune microenvironment is required.