Learn More
Selleck Chemical LLC Alantolactone
SDP
Catalog No. 5000000842
Encompass_Preferred
Alantolactone a naturally occurring eudesmane-type sesquiterpene lactone (SL) could induce activin/SMAD3 signaling and disrupt Cripto-1/activin receptor type II A interaction
Catalog No. 50-000-00842 Supplier Selleck Chemical LLC Supplier No. S831850MG
Alantolactone a naturally occurring eudesmane-type sesquiterpene lactone (SL) could induce activin/SMAD3 signaling and disrupt Cripto-1/activin receptor type II A interaction