Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alantolactone, Tocris Bioscience™

Catalog No. 449010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-901-0 10 mg
44-905-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-901-0 Supplier R&D Systems Supplier No. 4490/10
This item is not returnable. View return policy

Antiproliferative; induces activin/Smad3 signaling

Sesquiterpene lactone. Induces activin/SMAD3 signaling and disrupts Cripto-1/activin receptor type II A interaction. Displays antiproliferative activity in human colon adenocarcinoma HCT-8 cells.

Chemical Identifiers

CAS 546-43-0
Molecular Formula C15H20O2
Molecular Weight (g/mol) 232.32
MDL Number MFCD00274568
InChI Key PXOYOCNNSUAQNS-AGNJHWRGSA-N
Synonym alantolactone, helenine, eupatal, alant camphor, inula camphor, helenin, elecampane camphor, unii-m7gsn5q1m6, m7gsn5q1m6, 3ar,5s,8ar,9ar-5,8a-dimethyl-3-methylidene-3a,5,6,7,8,8a,9,9a-octahydronaphtho 2,3-b furan-2 3h-one
PubChem CID 72724
ChEBI CHEBI:2540
IUPAC Name (3aR,5S,8aR,9aR)-5,8a-dimethyl-3-methylidene-2H,3H,3aH,5H,6H,7H,8H,8aH,9H,9aH-naphtho[2,3-b]furan-2-one
SMILES C[C@H]1CCC[C@]2(C)C[C@H]3OC(=O)C(=C)[C@H]3C=C12

Specifications

Quantity 10 mg
Formula Weight 232.32
Percent Purity >98%
Chemical Name or Material Alantolactone
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.