Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alantolactone, Tocris Bioscience™
GREENER_CHOICE

Catalog No. 449050
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 44-905-0

Supplier: R&D Systems 4490/50

This item is not returnable. View return policy

Antiproliferative; induces activin/Smad3 signaling

Sesquiterpene lactone. Induces activin/SMAD3 signaling and disrupts Cripto-1/activin receptor type II A interaction. Displays antiproliferative activity in human colon adenocarcinoma HCT-8 cells.

Chemical Identifiers

CAS 546-43-0
Molecular Formula C15H20O2
Molecular Weight (g/mol) 232.32
MDL Number MFCD00274568
InChI Key PXOYOCNNSUAQNS-AGNJHWRGSA-N
Synonym alantolactone, helenine, eupatal, alant camphor, inula camphor, helenin, elecampane camphor, unii-m7gsn5q1m6, m7gsn5q1m6, 3ar,5s,8ar,9ar-5,8a-dimethyl-3-methylidene-3a,5,6,7,8,8a,9,9a-octahydronaphtho 2,3-b furan-2 3h-one
PubChem CID 72724
ChEBI CHEBI:2540
IUPAC Name (3aR,5S,8aR,9aR)-5,8a-dimethyl-3-methylidene-2H,3H,3aH,5H,6H,7H,8H,8aH,9H,9aH-naphtho[2,3-b]furan-2-one
SMILES C[C@H]1CCC[C@]2(C)C[C@H]3OC(=O)C(=C)[C@H]3C=C12

Specifications

Quantity 50 mg
Formula Weight 232.32
Percent Purity >98%
Chemical Name or Material Alantolactone
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.