Learn More
SPS Medical ALANTOR CUBE TEST PACK
Catalog No. NC9492363
Encompass
Lantor Cube Reusable Test Pack,25 tests/pk ,The industry's first and only reusable prevacuum sterilizer residual air test is comprised of 2 blocks of spun-bonded polypropylene placed on either side of a chemical indicator sheet
Catalog No. NC9492363 Supplier SPS Medical Supplier No. LCR025
Lantor Cube Reusable Test Pack,25 tests/pk ,The industry's first and only reusable prevacuum sterilizer residual air test is comprised of 2 blocks of spun-bonded polypropylene placed on either side of a chemical indicator sheet