Learn More
Apexbio Technology LLC Alarelin Acetate 79561-22-1 1g
SDP
Catalog No. 5000064097
Encompass_Preferred
Alarelin Acetate (CAS 79561-22-1) is a synthetic gonadotropin-releasing hormone (GnRH) agonist peptide This compound acts at the hypothalamic-pituitary axis by stimulating GnRH receptors leading to increased release of follicle-stimulating hormone (FSH) and luteinizing hormone (LH) from the anterior pituitary gland Alarelin demonstrates enhanced potency relative to natural LH-RH in stimulating gonadotropin secretion both in vivo and in vitro facilitating the induction of ovulation in animal models Given these endocrine-stimulating properties researchers commonly use Alarelin Acetate in studies focused on reproductive physiology fertility treatments and therapeutic interventions targeting endometriosis
Catalog No. 50-000-64097 Supplier Apexbio Technology LLC Supplier No. A31511000
Alarelin Acetate (CAS 79561-22-1) is a synthetic gonadotropin-releasing hormone (GnRH) agonist peptide This compound acts at the hypothalamic-pituitary axis by stimulating GnRH receptors leading to increased release of follicle-stimulating hormone (FSH) and luteinizing hormone (LH) from the anterior pituitary gland Alarelin demonstrates enhanced potency relative to natural LH-RH in stimulating gonadotropin secretion both in vivo and in vitro facilitating the induction of ovulation in animal models Given these endocrine-stimulating properties researchers commonly use Alarelin Acetate in studies focused on reproductive physiology fertility treatments and therapeutic interventions targeting endometriosis