Learn More
TARGETMOL CHEMICALS INC Alarelin Acetate 25MG
SDP
Catalog No. 502878248
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Alarelin Acetate is a synthetic GnRH agonist which can directly inhibit proliferation and DNA synthesis of rat GSMC through GnRH receptors.
Catalog No. 50-287-8248 Supplier TARGETMOL CHEMICALS INC Supplier No. T149425MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Alarelin Acetate is a synthetic GnRH agonist which can directly inhibit proliferation and DNA synthesis of rat GSMC through GnRH receptors.