Learn More
Medchemexpress LLC Alarelin Acetate | 79561-22-1 | 99.7% | 1287.42 | 5 MG
SDP
Catalog No. 5000386085
Encompass_Preferred
Alarelin Acetate is a synthetic GnRH agonist. It significantly lowers cell viability and achieves a maximum stimulatory effect on cell viability at a concentration of 10-5 M, acting in a dose-dependent manner. It can inhibit gastric acid secretion by directly acting on parietal cells in rats and by inhibiting vagous function, and can also significantly enhance the ratio of the G1 phase and decrease the ratio of the S phase of GSMC of rats.
  • Synthetic GnRH agonist.
  • Lowers cell viability significantly.
  • Inhibits gastric acid secretion.
  • Enhances G1 phase and decreases S phase in rat GSMC.
  • For research use only.
  • Not sold to patients.
Catalog No. 50-003-86085 Supplier Medchemexpress LLC Supplier No. HY174055MG
Alarelin Acetate is a synthetic GnRH agonist. It significantly lowers cell viability and achieves a maximum stimulatory effect on cell viability at a concentration of 10-5 M, acting in a dose-dependent manner. It can inhibit gastric acid secretion by directly acting on parietal cells in rats and by inhibiting vagous function, and can also significantly enhance the ratio of the G1 phase and decrease the ratio of the S phase of GSMC of rats.
  • Synthetic GnRH agonist.
  • Lowers cell viability significantly.
  • Inhibits gastric acid secretion.
  • Enhances G1 phase and decreases S phase in rat GSMC.
  • For research use only.
  • Not sold to patients.