Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alarelin acetate | 79561-22-1 | MFCD00167542 | 1287.42 | C60H86N16O16 | 10 MG
SDP

Catalog No. 5000298752
Encompass_Preferred

Alarelin acetate is a synthetic gonadotropin-releasing hormone (GnRH) agonist peptide supplied as a dry powder for research applications. It targets the GnRH receptor and is intended for laboratory and preclinical use; not for human or clinical administration.

  • Synthetic gonadotropin-releasing hormone (GnRH) agonist peptide.
  • Provided as a powder suitable for storage and formulation.
  • CAS number 79561-22-1 for unambiguous identification.
  • Molecular weight 1287.42 g/mol (manufacturer reported).
  • Storage: sealed, away from moisture; powder -80°C for 2 years or -20°C for 1 year.
  • Intended for research use only; not for clinical or therapeutic use.

Catalog No. 50-002-98752 Supplier Medchemexpress LLC Supplier No. HY1740510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alarelin acetate is a synthetic gonadotropin-releasing hormone (GnRH) agonist peptide supplied as a dry powder for research applications. It targets the GnRH receptor and is intended for laboratory and preclinical use; not for human or clinical administration.

  • Synthetic gonadotropin-releasing hormone (GnRH) agonist peptide.
  • Provided as a powder suitable for storage and formulation.
  • CAS number 79561-22-1 for unambiguous identification.
  • Molecular weight 1287.42 g/mol (manufacturer reported).
  • Storage: sealed, away from moisture; powder -80°C for 2 years or -20°C for 1 year.
  • Intended for research use only; not for clinical or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.