Learn More
Cayman Chemical Alarelin (acetate), Purity: ≥98%, CAS: 79561-22-1, 50 mg
SDP
Catalog No. 502989684
Encompass_Preferred
A peptide agonist of GnRH; binds to GnRHR (Kd = 0.2 nM); decreases GnRH and GnRHR expression in the preoptic area and pMBH, respectively, of ovariectomized estradiol-treated rats at 6 ng, i.c.v., indicating an ultrashort negative feedback loop; induces desensitization in the pituitary and decreases serum estradiol levels and ovarian and uterine weight in neonatal and adult female rats; reduces tumor growth in an HEC-1-B mouse xenograft model at 20, 40, and 80 µg/mg
Catalog No. 50-298-9684 Supplier Cayman Chemical Supplier No. 3099350 MG
A peptide agonist of GnRH; binds to GnRHR (Kd = 0.2 nM); decreases GnRH and GnRHR expression in the preoptic area and pMBH, respectively, of ovariectomized estradiol-treated rats at 6 ng, i.c.v., indicating an ultrashort negative feedback loop; induces desensitization in the pituitary and decreases serum estradiol levels and ovarian and uterine weight in neonatal and adult female rats; reduces tumor growth in an HEC-1-B mouse xenograft model at 20, 40, and 80 µg/mg