Learn More
Aviva Systems Biology Corp ALAS2 Antibody - N-terminal region (ARP41891_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41891_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ALAS2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ALAS2. Alternate names: ANH1, ASB, PRO2399, XLSA, ALASE, XLDPP, XLEPP, ALAS-EApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58035