Learn More
Aviva Systems Biology Corp ALAS2 Antibody - C-terminal region (ARP41889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALAS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ALAS2. Alternate names: ANH1, ASB, XLSA, ALASE, XLDPP, XLEPP, ALAS-EApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58034