Learn More
Medchemexpress LLC HY-111398 5mg Medchemexpress, ALB-127158(a) CAS:1173154-32-9 Purity:>98%
SDP
Catalog No. 501968856
Encompass_Preferred
Medchemexpress, HY-111398 5mg ALB-127158(a) CAS:1173154-32-9 ALB-127158(a) is a potent and selective melanin concentrating hormone 1 (MCH1) receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8856 Supplier Medchemexpress LLC Supplier No. HY1113985MG
Medchemexpress, HY-111398 5mg ALB-127158(a) CAS:1173154-32-9 ALB-127158(a) is a potent and selective melanin concentrating hormone 1 (MCH1) receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.