Learn More
Aviva Systems Biology Corp ALB Antibody - middle region (ARP41746_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41746_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ALB. Alternate names: DKFZp779N1935, PRO0883, PRO0903, PRO1341ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-57979