Learn More
U.S. Pharmacopeia Albendazole, 54965-21-8, MFCD00083232, 200 mg
Catalog No. NC1444657
Encompass
Empirical Formula (Hill Notation): C12H15N3O2S, Molecular Weight: 265.33, Synonym: Methyl 5-(propylthio)-2-benzimidazolecarbamate.
Catalog No. NC1444657 Supplier U.S. Pharmacopeia Supplier No. 1012553
Empirical Formula (Hill Notation): C12H15N3O2S, Molecular Weight: 265.33, Synonym: Methyl 5-(propylthio)-2-benzimidazolecarbamate.