Learn More
Selleck Chemical LLC Albendazole S1640-1g
SDP
Catalog No. 503108654
Encompass_Preferred
Albendazole is a member of the benzimidazole compounds used as a drug indicated for the treatment of a variety of worm infestations
Catalog No. 50-310-8654 Supplier Selleck Chemical LLC Supplier No. S16401G
Albendazole is a member of the benzimidazole compounds used as a drug indicated for the treatment of a variety of worm infestations