Learn More
Apexbio Technology LLC Albendazole 54965-21-8 100mg
SDP
Catalog No. 5000096194
Encompass_Preferred
Albendazole (CAS 54965-21-8) is a small-molecule inhibitor targeting tubulin It is designed to disrupt microtubule assembly thereby impairing cellular cytoskeletal function and parasite viability Albendazole exerts its biological activity primarily through binding to tubulin and inhibiting microtubule polymerization In tumor cell-based assays Albendazole demonstrates antiproliferative activity and induces cell cycle arrest autophagy and apoptosis with reported IC50 values typically ranging from 0 1 to 1 M depending on cell line and experimental conditions Based on these pharmacological properties Albendazole holds research potential in studies of parasitic infections microtubule dynamics tumor cell proliferation angiogenesis and programmed cell death
Catalog No. 50-000-96194 Supplier Apexbio Technology LLC Supplier No. A8358100
Add to Cart
Add to Cart
Albendazole (CAS 54965-21-8) is a small-molecule inhibitor targeting tubulin It is designed to disrupt microtubule assembly thereby impairing cellular cytoskeletal function and parasite viability Albendazole exerts its biological activity primarily through binding to tubulin and inhibiting microtubule polymerization In tumor cell-based assays Albendazole demonstrates antiproliferative activity and induces cell cycle arrest autophagy and apoptosis with reported IC50 values typically ranging from 0 1 to 1 M depending on cell line and experimental conditions Based on these pharmacological properties Albendazole holds research potential in studies of parasitic infections microtubule dynamics tumor cell proliferation angiogenesis and programmed cell death