Learn More
Medchemexpress LLC Albendazole | 54965-21-8 | 99.1% | 1 ML
SDP
Catalog No. 5000406367
Encompass_Preferred
Albendazole is an orally active, broad-spectrum parasiticide used for research on gastrointestinal parasites in humans and animals. It is characterized by high effectiveness and low host toxicity, and has been shown to induce apoptosis and autophagy in various cancer cells. This compound also inhibits tubulin polymerization, HIF-1α, and VEGF expression, offering antioxidant activity and inhibiting the glycolytic process in cancer cells.
  • Broad-spectrum parasiticide for human and animal research
  • Induces apoptosis and autophagy in cancer cells
  • Inhibits tubulin polymerization
  • Offers antioxidant activity
  • Inhibits HIF-1α and VEGF expression
  • Inhibits the glycolytic process in cancer cells
Catalog No. 50-004-06367 Supplier Medchemexpress LLC Supplier No. HYB022310MM/1ML
Albendazole is an orally active, broad-spectrum parasiticide used for research on gastrointestinal parasites in humans and animals. It is characterized by high effectiveness and low host toxicity, and has been shown to induce apoptosis and autophagy in various cancer cells. This compound also inhibits tubulin polymerization, HIF-1α, and VEGF expression, offering antioxidant activity and inhibiting the glycolytic process in cancer cells.
  • Broad-spectrum parasiticide for human and animal research
  • Induces apoptosis and autophagy in cancer cells
  • Inhibits tubulin polymerization
  • Offers antioxidant activity
  • Inhibits HIF-1α and VEGF expression
  • Inhibits the glycolytic process in cancer cells