Learn More
Medchemexpress LLC Albendazole-d3 | 1353867-92-1 | 98.3% | 268.35 | C12H12D3N3O2S | 10 MG
SDP
Catalog No. 5000297092
Encompass_Preferred
Albendazole-d3 is the deuterium-labeled analogue of albendazole supplied as a research-grade reference standard. It is intended for use as a stable-isotope internal standard or tracer in quantitative analyses and is provided as a powder for analytical applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity for improved analytical accuracy.
  • Supplied as a 10 mg powder for trace-level assays.
  • Stable when stored at recommended temperatures.
Catalog No. 50-002-97092 Supplier Medchemexpress LLC Supplier No. HYB0223S10MG
Albendazole-d3 is the deuterium-labeled analogue of albendazole supplied as a research-grade reference standard. It is intended for use as a stable-isotope internal standard or tracer in quantitative analyses and is provided as a powder for analytical applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity for improved analytical accuracy.
  • Supplied as a 10 mg powder for trace-level assays.
  • Stable when stored at recommended temperatures.