Learn More
Medchemexpress LLC Albendazole-d3 (SKF-62979-d3) | 1353867-92-1 | 98.3% | 1 MG
SDP
Catalog No. 5000406233
Encompass_Preferred
Albendazole-d3 is the deuterium labeled Albendazole, a benzimidazole compound used as an agent for treating various worm infestations. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06233 Supplier Medchemexpress LLC Supplier No. HYB0223S1MG
Albendazole-d3 is the deuterium labeled Albendazole, a benzimidazole compound used as an agent for treating various worm infestations. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS