Learn More
Abcam Albendazole, Microtubule depolymerizing agent, 10G
Catalog No. 502706769
Encompass_Preferred
MW 265.33 Da, Purity >98%. Anthelmintic benzimidazole and microtubule depolymerizing agent. Able to inhibit brain microtubule polymerization in vitro and delays microtubule assembly in vivo. Anti-tumour activity seen in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6769 Supplier Abcam Supplier No. AB14124110G
MW 265.33 Da, Purity >98%. Anthelmintic benzimidazole and microtubule depolymerizing agent. Able to inhibit brain microtubule polymerization in vitro and delays microtubule assembly in vivo. Anti-tumour activity seen in vivo.
The product is subject to the following: Abcam Restricted Use Statement