Learn More
Apexbio Technology LLC Albendazole Oxide 54029-12-8 50mg
SDP
Catalog No. 5000089906
Encompass_Preferred
Albendazole Oxide (CAS 54029-12-8) is a small-molecule inhibitor targeting beta-tubulin It is designed to inhibit beta-tubulin polymerization in parasites thereby disrupting cytoplasmic microtubule structure and interfering with glucose metabolic pathways Albendazole Oxide exerts its biological activity primarily through inhibition of beta-tubulin polymerization In both in vitro and in vivo studies Albendazole Oxide demonstrates anti-parasitic activity with reported IC50 values typically ranging from nanomolar to micromolar concentrations depending on the parasite species and assay system Based on these pharmacological properties Albendazole Oxide holds research potential in the exploration of anti-parasitic pharmacodynamics notably for studies involving Echinococcus spp and other parasitic infections
Catalog No. 50-000-89906 Supplier Apexbio Technology LLC Supplier No. B188050
Albendazole Oxide (CAS 54029-12-8) is a small-molecule inhibitor targeting beta-tubulin It is designed to inhibit beta-tubulin polymerization in parasites thereby disrupting cytoplasmic microtubule structure and interfering with glucose metabolic pathways Albendazole Oxide exerts its biological activity primarily through inhibition of beta-tubulin polymerization In both in vitro and in vivo studies Albendazole Oxide demonstrates anti-parasitic activity with reported IC50 values typically ranging from nanomolar to micromolar concentrations depending on the parasite species and assay system Based on these pharmacological properties Albendazole Oxide holds research potential in the exploration of anti-parasitic pharmacodynamics notably for studies involving Echinococcus spp and other parasitic infections