Learn More
Selleck Chemical LLC Albendazole Oxide S1836-50mg
SDP
Catalog No. 503110747
Encompass_Preferred
Albendazole Oxide (Ricobendazole Albendazole sulfoxide) is a tubulin polymerization or assembly inhibitor used for the treatment of a variety of parasitic worm infestations
Catalog No. 50-311-0747 Supplier Selleck Chemical LLC Supplier No. S183650MG
Albendazole Oxide (Ricobendazole Albendazole sulfoxide) is a tubulin polymerization or assembly inhibitor used for the treatment of a variety of parasitic worm infestations