Learn More
Medchemexpress LLC Albendazole (standard) | 54965-21-8 | 98.3% | 100 MG
SDP
Catalog No. 5000406121
Encompass_Preferred
Albendazole (Standard) is the analytical standard of Albendazole, intended for research and analytical applications. Albendazole is an orally active and broad-spectrum parasiticide with high effectiveness and low host toxicity, used for the research of gastrointestinal parasites in humans and animals. This compound is a grade of analytical standard, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Induces apoptosis and autophagy in cancer cells
  • Inhibits tubulin polymerization and HIF-1α, VEGF expression
  • Possesses antioxidant activity
  • Inhibits the glycolytic process in cancer cells
Catalog No. 50-004-06121 Supplier Medchemexpress LLC Supplier No. HYB0223R100MG
Albendazole (Standard) is the analytical standard of Albendazole, intended for research and analytical applications. Albendazole is an orally active and broad-spectrum parasiticide with high effectiveness and low host toxicity, used for the research of gastrointestinal parasites in humans and animals. This compound is a grade of analytical standard, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Induces apoptosis and autophagy in cancer cells
  • Inhibits tubulin polymerization and HIF-1α, VEGF expression
  • Possesses antioxidant activity
  • Inhibits the glycolytic process in cancer cells