Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Albendazole (standard) | 54965-21-8 | 98.3% | 265.33 | C12H15N3O2S | 10 MG
SDP

Catalog No. 5000298263
Encompass_Preferred

Albendazole (standard) is an analytical reference standard of the anthelmintic compound albendazole intended for laboratory and analytical use. Supplied as a certified standard suitable for method development, quantitative assays, and stability testing.

  • Purity: 98.3%.
  • Molecular weight: 265.33 g/mol.
  • Chemical formula: C12H15N3O2S.
  • CAS number: 54965-21-8.
  • Pack size: 10 mg (additional sizes available).
  • Documentation: COA and SDS available for batch verification.

Catalog No. 50-002-98263 Supplier Medchemexpress LLC Supplier No. HYB0223R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Albendazole (standard) is an analytical reference standard of the anthelmintic compound albendazole intended for laboratory and analytical use. Supplied as a certified standard suitable for method development, quantitative assays, and stability testing.

  • Purity: 98.3%.
  • Molecular weight: 265.33 g/mol.
  • Chemical formula: C12H15N3O2S.
  • CAS number: 54965-21-8.
  • Pack size: 10 mg (additional sizes available).
  • Documentation: COA and SDS available for batch verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.