Learn More
Medchemexpress LLC Albendazole (Standard) | 54965-21-8 | 98.3% | 50 MG
SDP
Catalog No. 5000405435
Encompass_Preferred
Albendazole (Standard) is the analytical standard of Albendazole, intended for research and analytical applications. This orally active, broad-spectrum parasiticide is known for its high effectiveness and low host toxicity, and is utilized in the research of gastrointestinal parasites in both humans and animals.
  • Induces apoptosis and autophagy in cancer cells.
  • Inhibits tubulin polymerization.
  • Inhibits HIF-1α and VEGF expression.
  • Possesses antioxidant activity.
  • Hinders the glycolytic process in cancer cells.
  • Commonly used in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Serves as a reference standard for assays.
Catalog No. 50-004-05435 Supplier Medchemexpress LLC Supplier No. HYB0223R50MG
Add to Cart
Add to Cart
Albendazole (Standard) is the analytical standard of Albendazole, intended for research and analytical applications. This orally active, broad-spectrum parasiticide is known for its high effectiveness and low host toxicity, and is utilized in the research of gastrointestinal parasites in both humans and animals.
  • Induces apoptosis and autophagy in cancer cells.
  • Inhibits tubulin polymerization.
  • Inhibits HIF-1α and VEGF expression.
  • Possesses antioxidant activity.
  • Hinders the glycolytic process in cancer cells.
  • Commonly used in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Serves as a reference standard for assays.