Learn More
Medchemexpress LLC Albendazole (Standard) (SKF-62979 (Standard)) | 54965-21-8 | 98.3% | 25 MG
SDP
Catalog No. 5000450533
Encompass_Preferred
Albendazole (Standard) is an analytical standard for research and analytical applications. This orally active, broad-spectrum parasiticide shows high effectiveness and low host toxicity, used for studying gastrointestinal parasites. It induces apoptosis and autophagy in cancer cells, inhibits tubulin polymerization, HIF-1α, and VEGF expression, and demonstrates antioxidant activity, inhibiting the glycolytic process in cancer cells.
  • Analytical standard for research and analysis
  • Orally active, broad-spectrum parasiticide
  • High effectiveness and low host toxicity
  • Used for research of gastrointestinal parasites
  • Induces apoptosis and autophagy in cancer cells
  • Inhibits tubulin polymerization, HIF-1α, and VEGF expression
  • Exhibits antioxidant activity
  • Inhibits glycolytic process in cancer cells
  • Applied in HPLC, GC, and MS research experiments
Catalog No. 50-004-50533 Supplier Medchemexpress LLC Supplier No. 25MGHYB0223R
Albendazole (Standard) is an analytical standard for research and analytical applications. This orally active, broad-spectrum parasiticide shows high effectiveness and low host toxicity, used for studying gastrointestinal parasites. It induces apoptosis and autophagy in cancer cells, inhibits tubulin polymerization, HIF-1α, and VEGF expression, and demonstrates antioxidant activity, inhibiting the glycolytic process in cancer cells.
  • Analytical standard for research and analysis
  • Orally active, broad-spectrum parasiticide
  • High effectiveness and low host toxicity
  • Used for research of gastrointestinal parasites
  • Induces apoptosis and autophagy in cancer cells
  • Inhibits tubulin polymerization, HIF-1α, and VEGF expression
  • Exhibits antioxidant activity
  • Inhibits glycolytic process in cancer cells
  • Applied in HPLC, GC, and MS research experiments