Learn More
Cayman Chemical Albendazole sulfone, Purity: ≥98%, CAS: 75184-71-3, 10 mg
SDP
Catalog No. 502913564
Encompass_Preferred
An active metabolite of albendazole; inhibits spore formation of E. hellem, E. cuniculi, and E. intestinalis in vitro (IC90s = <0.0001, <0.001, and <0.1 µg/ml, respectively)
Catalog No. 50-291-3564 Supplier Cayman Chemical Supplier No. 3544510 MG
An active metabolite of albendazole; inhibits spore formation of E. hellem, E. cuniculi, and E. intestinalis in vitro (IC90s = <0.0001, <0.001, and <0.1 µg/ml, respectively)