Learn More
Cayman Chemical Albendazole sulfone-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1448345-60-5, 1 mg
SDP
Catalog No. 502910504
Encompass_Preferred
An internal standard for the quantification of albendazole sulfone by GC- or LC-MS
Catalog No. 50-291-0504 Supplier Cayman Chemical Supplier No. 351291 MG
An internal standard for the quantification of albendazole sulfone by GC- or LC-MS