Learn More
TARGETMOL CHEMICALS INC Albendazole sulfoxide 500MG
SDP
Catalog No. 502877066
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Albendazole sulfoxide (Ricobendazole) is a metabolite of Albendazole, an anthelmintic. Purity 98.04%
Catalog No. 50-287-7066 Supplier TARGETMOL CHEMICALS INC Supplier No. T6372500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Albendazole sulfoxide (Ricobendazole) is a metabolite of Albendazole, an anthelmintic. Purity 98.04%