Learn More
Medchemexpress LLC Albendazole sulfoxide | 54029-12-8 | 99.2% | 1 MG
SDP
Catalog No. 5000342205
Encompass_Preferred
Albendazole sulfoxide is the main active metabolite of Albendazole, showing an anti-parasite effect against *Echinococcus multilocularis Metacestodes*. It is for research use only.
  • Exhibits parasitocidal effect against *E. multilocularis* metacestodes in vitro.
  • Reduces cyst size and weight in Balb/c mice infected with protoscoleces in vivo.
  • Available in various forms including deuterated versions.
  • Provided with comprehensive documentation and technical support.
Catalog No. 50-003-42205 Supplier Medchemexpress LLC Supplier No. HY12785S1MG
Albendazole sulfoxide is the main active metabolite of Albendazole, showing an anti-parasite effect against *Echinococcus multilocularis Metacestodes*. It is for research use only.
  • Exhibits parasitocidal effect against *E. multilocularis* metacestodes in vitro.
  • Reduces cyst size and weight in Balb/c mice infected with protoscoleces in vivo.
  • Available in various forms including deuterated versions.
  • Provided with comprehensive documentation and technical support.